Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CXCL17 Monoclonal Antibody (422208)

Mouse Monoclonal Antibody

Supplier:  Invitrogen™ MA524157

Catalog No. PIMA524157


Only null left
Add to Cart

Description

Description

In direct ELISAs and Western blots, approximately 25% cross-reactivity with recombinant mouse CXCL17 is observed. Reconstitute at 0.5 mg/mL in sterile PBS.

CXCL17 is a chemokine that acts as chemoattractant for monocytes, macrophages and dendritic cells. In particular, CXCL17 is a mucosal chemokine that attracts dendritic cells and blood monocytes to the lungs. CXCL17 plays a role in angiogenesis and possibly in the development of tumors. Further, CXCL17 acts as an anti-inflammatory in the stomach and may play a role in the innate defense against infections. CXCL17 also activates the C-X-C chemokine receptor GPR35 to induce a rapid and transient rise in the level of intracellular calcium ions. 4-Cys CXCL17 seems to exhibit much higher chemoattractant potency on monocytes and macrophages than 6-Cys CXCL17. Diseases associated with CXCL17 dysfunction include cholera.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

CXCL17
Monoclonal
0.5 mg/mL
PBS with 5% trehalose and No Preservative
Q6UXB2
CXCL17
E. coli-derived recombinant human CXCL17/VCC-1 Leu24-Leu119.
100 μg
Primary
Human
Antibody
IgG2b
Flow Cytometry, Western Blot, Immunocytochemistry
422208
Unconjugated
CXCL17
13.6 kDa protein; 13.6-kDa protein; 4-Cys CXCL17; 6-Cys CXCL17; C Cmotif chemokine; C X C motif chemokine; CC motif chemokine; CCmotif chemokine; chemokine (C-X-C motif) ligand 17; CXC; CXC motif chemokine; C-X-C motif chemokine 17; C-X-C motif chemokine ligand 17; CXCL; CXCL17; Dcip1; Dendritic cell and monocyte chemokine-like protein; DMC; UNQ473; UNQ473/PRO842; Vcc1; VCC-1; VEGF coregulated chemokine 1; VEGF co-regulated chemokine 1
Mouse
Protein A/G
RUO
284340
-20°C, Avoid Freeze/Thaw Cycles
Lyophilized
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.