Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CXCL17/VCC-1 Antibody (422208), HRP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB4207H
Description
CXCL17/VCC-1 Monoclonal antibody specifically detects CXCL17/VCC-1 in Human samples. It is validated for Western Blot, Flow Cytometry (Intracellular Staining), Immunocytochemistry, CyTOFSpecifications
CXCL17/VCC-1 | |
Monoclonal | |
HRP | |
PBS | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot, Flow Cytometry (Intracellular Staining), Immunocytochemistry, CyTOF | |
422208 | |
Western Blot, Intracellular Staining by Flow Cytometry, Immunocytochemistry, CyTOF-ready | |
Chemokine (C-X-C Motif) Ligand 17, C-X-C Motif Chemokine 17, Dcip1, Dendritic Cell And Monocyte Chemokine-Like Protein, DMC, UNQ473, VCC1, VCC-1, VEGF Coregulated Chemokine 1, VEGF Co-Regulated Chemokine 1 | |
E. coli-derived recombinant human CXCL17/VCC-1, Leu24-Leu119, Accession # Q6UXB2 | |
0.1 mL | |
Biologically Active Proteins | |
284340 | |
Store at 4°C in the dark. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction