Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CXCL2/GRO beta/MIP-2/CINC-3 Antibody (126), FITC, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290860F
Description
CXCL2/GRO beta/MIP-2/CINC-3 Monoclonal specifically detects CXCL2/GRO beta/MIP-2/CINC-3 in Rat samples. It is validated for ELISA.Specifications
CXCL2/GRO beta/MIP-2/CINC-3 | |
Monoclonal | |
FITC | |
chemokine (C-X-C motif) ligand 2, CINC-2a, C-X-C Motif Chemokine 2, GRO2, GRO2 oncogene, GROb, Gro-beta, Growth-regulated protein beta, Macrophage inflammatory protein 2-alpha, melanoma growth stimulatory activity beta, MGSA beta, MGSA-b, MIP2, MIP-2a, MIP2A, MIP2-alpha, SCYB2 | |
This antibody was obtained from a rabbit immunized with purified, recombinant Rat CXCL2/GRO beta/MIP-2/CINC-3 (NP_446099.1; Ser32-Asn100). | |
0.1 mL | |
Biologically Active Proteins, Chemokines and Cytokines, Signal Transduction | |
2920 | |
Store at 4°C in the dark. | |
IgG |
ELISA | |
126 | |
PBS | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction