Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CXCL2/GRO beta/MIP-2/CINC-3 Antibody, mFluor Violet 610 SE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP299400MFV610
Description
CXCL2/GRO beta/MIP-2/CINC-3 Polyclonal antibody specifically detects CXCL2/GRO beta/MIP-2/CINC-3 in Human samples. It is validated for ELISASpecifications
CXCL2/GRO beta/MIP-2/CINC-3 | |
Polyclonal | |
ELISA | |
chemokine (C-X-C motif) ligand 2, CINC-2a, C-X-C Motif Chemokine 2, GRO2, GRO2 oncogene, GROb, Gro-beta, Growth-regulated protein beta, Macrophage inflammatory protein 2-alpha, melanoma growth stimulatory activity beta, MGSA beta, MGSA-b, MIP2, MIP-2a, MIP2A, MIP2-alpha, SCYB2 | |
Produced in rabbits immunized with purified, recombinant Human CXCL2/GRO beta/MIP-2/CINC-3 (Accession#: NP_002080.1; Ala35-Asn107) | |
0.1 mL | |
Biologically Active Proteins, Chemokines and Cytokines, Signal Transduction | |
2920 | |
Store at 4C in the dark. | |
IgG |
ELISA | |
mFluor Violet 610 SE | |
50mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction