Learn More
CXCL2 Polyclonal Antibody, Biotin, PeproTech®, Invitrogen™
Rabbit Polyclonal Antibody
$250.00 - $2175.00
Specifications
Antigen | CXCL2 |
---|---|
Concentration | 0.1-1.0 mg/mL |
Applications | ELISA, Western Blot |
Classification | Polyclonal |
Conjugate | Biotin |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
500P75BT25
![]() |
Invitrogen™
500P75BT25UG |
25 μg |
Each for $250.00
|
|
|||||
500P75BT50
![]() |
Invitrogen™
500P75BT50UG |
50 μg |
Each for $319.00
|
|
|||||
50-271-8452
![]() |
Invitrogen™
500P75BT500UG |
500 μg |
Each for $2,175.00
|
|
|||||
Description
AA Sequence of recombinant protein: VVVASELRCQ CLTTLPRVDF KNIQSLTVTP PGPHCAQTEV IATLKDGHEV CLNPEAPLVQ RIVQKILNKG KAN. Preparation: Produced from sera of rabbits immunized with highly pure Recombinant Rat GRO-beta/MIP-2 (CXCL2). Anti-Rat GRO-beta/MIP-2 (CXCL2)-specific antibody was purified by affinity chromatography and then biotinylated. Sandwich ELISA: To detect Rat GRO-beta/MIP-2 by sandwich ELISA (using 100 μL/well antibody solution) a concentration of 0.25-1.0 μg/mL of this antibody is required. This biotinylated polyclonal antibody, in conjunction with PeproTech Polyclonal Anti-Rat GRO-beta/MIP-2 (500-P75) as a capture antibody, allows the detection of at least 0.2-0.4 ng/well of Recombinant Rat GRO-beta/MIP-2. Western Blot: To detect Rat GRO-beta/MIP-2 by Western Blot analysis this antibody can be used at a concentration of 0.1-0.2 μg/mL. Used in conjunction with compatible secondary reagents the detection limit for Recombinant Rat GRO-beta/MIP-2 is 1.5-3.0 ng/lane, under either reducing or non-reducing conditions. 500-P75BT-1MG will be provided as 2 x 500 μg
Produced by activated monocytes and neutrophils and expressed at sites of inflammation. Hematoregulatory chemokine, which, in vitro, suppresses hematopoietic progenitor cell proliferation. GRO-beta(5-73) shows a highly enhanced hematopoietic activity.Specifications
CXCL2 | |
ELISA, Western Blot | |
Biotin | |
Rabbit | |
Rat | |
P30348 | |
114105 | |
E.coli-derived Recombinant Rat GRO-beta/MIP-2 (CXCL2). | |
Antigen affinity chromatography | |
Cxcl2 |
0.1-1.0 mg/mL | |
Polyclonal | |
Lyophilized | |
RUO | |
PBS with no preservative | |
C Cmotif chemokine; C X C motif chemokine; CC motif chemokine; CCmotif chemokine; chemokine (C-X-C motif) ligand 2; CINC-2a; Cinc3; CINC-3; CXC; CXC motif chemokine; C-X-C motif chemokine 2; C-X-C motif chemokine ligand 2; CXCL; CXCL2; Cytokine-induced neutrophil chemoattractant 3; GRO b; gro beta; GRO ß; GRO2; GRO2 oncogene; GROb; GRObeta; gro-beta; GRO-beta(5-73); GRO-beta-T; Growth-regulated protein beta; GROß; Hematopoietic synergistic factor; HSF; Macrophage inflammatory protein 2; macrophage inflammatory protein 2-alpha; melanoma growth stimulatory activity beta; MGSA beta; Mgsa-b; MIP 2 a; MIP 2 alpha; MIP 2 a; Mip2; MIP-2; MIP2 a; MIP2 a; MIP2A; MIP-2a; MIP2-alpha; SB-251353; Scyb; Scyb2; small inducible cytokine subfamily, member 2 | |
Cxcl2 | |
Primary | |
-20°C |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.