Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PeproTech CXCL2 Polyclonal Antibody, PeproTech®, Invitrogen™

Catalog No. 500P7550UG
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
100 μg
50 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
500P7550UG 50 μg
500P75100UG 100 μg
500-P75-1MG 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 500P7550UG Supplier PeproTech Supplier No. 500P7550UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

AA Sequence of recombinant protein: VVVASELRCQ CLTTLPRVDF KNIQSLTVTP PGPHCAQTEV IATLKDGHEV CLNPEAPLVQ RIVQKILNKG KAN. Preparation: Produced from sera of rabbits immunized with highly pure Recombinant Rat GRO-beta/MIP-2 (CXCL2). Anti-Rat GRO-beta/MIP-2 (CXCL2)-specific antibody was purified by affinity chromatography employing an immobilized Rat GRO-beta/MIP-2 (CXCL2) matrix. Sandwich ELISA: To detect Rat GRO-beta/MIP-2 (CXCL2) by sandwich ELISA (using 100 μL/well antibody solution) a concentration of 0.5-2.0 μg/mL of this antibody is required. This antigen affinity purified antibody, in conjunction with PeproTech Biotinylated Anti-Rat GRO-beta/MIP-2 (CXCL2) (500-P75Bt) as a detection antibody, allows the detection of at least 0.2-0.4 ng/well of Recombinant Rat GRO-beta/MIP-2 (CXCL2). Western Blot: To detect Rat GRObeta/MIP-2 by Western Blot analysis this antibody can be used at a concentration of 0.1-0.2 μg/mL. When used in conjunction with compatible secondary reagents, the detection limit for Recombinant Rat GRObeta/MIP-2 is 1.5-3.0 ng/lane, under either reducing or non-reducing conditions.

Produced by activated monocytes and neutrophils and expressed at sites of inflammation. Hematoregulatory chemokine, which, in vitro, suppresses hematopoietic progenitor cell proliferation. GRO-beta(5-73) shows a highly enhanced hematopoietic activity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CXCL2
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.1-1.0 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene Cxcl2
Gene Accession No. P30348
Gene Alias C Cmotif chemokine; C X C motif chemokine; CC motif chemokine; CCmotif chemokine; chemokine (C-X-C motif) ligand 2; CINC-2a; Cinc3; CINC-3; CXC; CXC motif chemokine; C-X-C motif chemokine 2; C-X-C motif chemokine ligand 2; CXCL; CXCL2; Cytokine-induced neutrophil chemoattractant 3; GRO b; gro beta; GRO β; GRO2; GRO2 oncogene; GROb; GRObeta; gro-beta; GRO-beta(5-73); GRO-beta-T; Growth-regulated protein beta; GROβ; Hematopoietic synergistic factor; HSF; Macrophage inflammatory protein 2; macrophage inflammatory protein 2-alpha; melanoma growth stimulatory activity beta; MGSA beta; Mgsa-b; MIP 2 a; MIP 2 alpha; MIP 2 α; Mip2; MIP-2; MIP2 a; MIP2 α; MIP2A; MIP-2a; MIP2-alpha; SB-251353; Scyb; Scyb2; small inducible cytokine subfamily, member 2
Gene Symbols Cxcl2
Host Species Rabbit
Immunogen E.coli-derived Recombinant Rat GRO-beta/MIP-2 (CXCL2).
Purification Method Antigen affinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 114105
Target Species Rat
Content And Storage -20°C
Product Type Antibody
Form Lyophilized
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.