Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CXCL3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5103136
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5103136 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5103136 Supplier Invitrogen™ Supplier No. PA5103136
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total GRO gamma.

CXCL3 may play a role in inflammation and exert its effects on endothelial cells in an autocrine fashion. All three isoforms of GRO are CXC chemokines that can signal through the CXCR1 or CXCR2 receptors. The GRO proteins chemoattract and activate neutrophils and basophils.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CXCL3
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene Cxcl3
Gene Accession No. P19876, Q10746, Q6W5C0
Gene Alias C Cmotif chemokine; C X C motif chemokine; CC motif chemokine; CCmotif chemokine; chemokine (C-X-C motif) ligand 3; Cinc2; CINC-2; CINC-2b; CXC; CXC motif chemokine; C-X-C motif chemokine 3; C-X-C motif chemokine ligand 3; CXCL; Cxcl3; cytokine-induced neutrophil chemoattractant 2; cytokine-induced neutrophil chemoattractant-2; Dcip1; Dendritic cell inflammatory protein 1; Gm1960; gro gamma; GRO3; GRO3 oncogene; GROG; GRO-gamma; GRO-gamma(1-73); GRO-gamma(5-73); Growth-regulated protein gamma; macrophage inflammatory protein 2-alpha/beta; Macrophage inflammatory protein 2-beta; melanoma growth stimulatory activity gamma; MGSA gamma; MIP2-alpha/beta; MIP2B; MIP-2b; MIP2-beta; SCYB3
Gene Symbols Cxcl3
Host Species Rabbit
Immunogen A synthesized peptide derived from human CXCL3(Accession P19876), corresponding to amino acid residues E73-N107.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 171551, 2921, 330122
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.