Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CXCL4/PF4 Rabbit anti-Human, Mouse, Rat, Clone: 6Q10H2, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP316251100UL
This item is not returnable.
View return policy
Description
CXCL4/PF4 Monoclonal antibody specifically detects CXCL4/PF4 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
CXCL4/PF4 | |
Monoclonal | |
Unconjugated | |
chemokine (C-X-C motif) ligand 4, C-X-C motif chemokine 4, CXCL4iroplact, Iroplact, MGC138298, Oncostatin-A, PF-4, platelet factor 4, SCYB4oncostatin-A | |
A synthetic peptide corresponding to a sequence within amino acids 1-101 of human CXCL4/CXCL4/PF4 (P02776). | |
100 μg | |
Biologically Active Proteins, Cancer, Cardiovascular Biology, Signal Transduction | |
5196 | |
Store at -20°C. Avoid freeze-thaw cycles. | |
IgG |
Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
6Q10H2 | |
PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction