Learn More
CXCL5/ENA-78 Antibody (32B3) - Azide and BSA Free, Novus Biologicals™
SDP
Rat Monoclonal Antibody
Supplier:  Novus Biologicals NBP222026
 View more versions of this product
Catalog No. NBP222026
Description
Description
CXCL5/ENA-78 Monoclonal specifically detects CXCL5/ENA-78 in Mouse samples. It is validated for Western Blot, ELISA, Block/Neutralize.
Specifications
Specifications
CXCL5
Monoclonal
LYOPH
Western Blot 1:1500, ELISA 1:1000-1:2000, Block/Neutralize
P50228
CXCL5
Mouse recombinant CXCL5/ENA-78
0.1 mg
Cell Cycle and Replication
6374
Mouse
Purified
Western Blot, ELISA, Blocking Assay
32B3
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS. 0.025 mg size is provided in liquid form, PBS with No Preservative
chemokine (C-X-C motif) ligand 5, C-X-C motif chemokine 5, ENA-78, ENA-78(1-78), epithelial-derived neutrophil activating protein 78, epithelial-derived neutrophil-activating peptide 78, Epithelial-derived neutrophil-activating protein 78, Neutrophil-activating peptide ENA-78, neutrophil-activating protein 78, SCYB5ENA78, small inducible cytokine B5, small inducible cytokine subfamily B (Cys-X-Cys), member 5 (epithelial-derivedneutrophil-activating peptide 78), Small-inducible cytokine B5
Rat
Protein G purified
RUO
Primary
Reconstitute with sterilized PBS to a final concentration of 0.2 mg/ml.
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
IgG2
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.
For Research Use Only