Learn More
CXCL5/ENA-78 Antibody (MM0216-10L19), Alexa Fluor™ 750, Novus Biologicals™
SDP
Mouse Monoclonal Antibody
Supplier:  Novus Biologicals NBP212231AF750
Catalog No. NP212231A70
Only null left
Add to Cart
Description
Description
CXCL5/ENA-78 Monoclonal specifically detects CXCL5/ENA-78 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
Specifications
CXCL5/ENA-78
Monoclonal
Alexa Fluor 750
50 mM sodium borate with 0.05% sodium azide
CXCL5
Human recombinant CXCL5/ENA-78
Primary
Human
IgG1
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
MM0216-10L19
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
chemokine (C-C motif) ligand 5, C-X-C motif chemokine 5, ENA-78, ENA-78(1-78), Epithelial-derived neutrophil-activating protein 78, Neutrophil-activating peptide ENA-78, neutrophil-activating protein 78, SCYB5ENA78
Mouse
0.1 mL
6374
Store at 4°C in the dark.
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.