Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CXCL5 Monoclonal Antibody (03), Invitrogen™

Catalog No. PIMA543387
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA543387 50 μL
PIMA543007 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. PIMA543387 Supplier Invitrogen™ Supplier No. MA543387
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Specificity: Human CXCL5

The protein encoded by this gene is an inflammatory chemokine that belongs to the CXC chemokine family. This chemokine is produced concomitantly with interleukin-8 (IL8) in response to stimulation with either interleukin-1 (IL1) or tumor necrosis factor-alpha (TNFA). This chemokine is a potent chemotaxin involved in neutrophil activation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CXCL5
Applications ELISA
Classification Monoclonal
Clone 03
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene CXCL5
Gene Accession No. P42830
Gene Alias AMCF-II; C Cmotif chemokine; C X C motif chemokine; CC motif chemokine; CCmotif chemokine; chemokine (C-X-C motif) ligand 5; chemokine (C-X-C motif) ligand 6; chemokine (C-X-C motif) ligand 6 (granulocyte chemotactic protein 2); CXC; CXC chemokine LIX; CXC motif chemokine; C-X-C motif chemokine 5; C-X-C motif chemokine ligand 5; CXCL; CXCL5; Cxcl6; Cytokine LIX; ENA78; ENA-78; ENA-78(1-78); ENA-78(8-78); ENA-78(9-78); Epithelial-derived neutrophil-activating protein 78; GCP-2; GCP-2(1-78); GCP-2(9-78); granulocyte chemotactic protein-2; LIX; Neutrophil-activating peptide ENA-78; neutrophil-activating protein 78; Scyb5; Scyb6; small inducible cytokine B subfamily, member 5; small inducible cytokine subfamily B (Cys-X-Cys), member 5 (epithelial-derived neutrophil-activating peptide 78); small inducible cytokine subfamily B, member 15; small-inducible cytokine B5
Gene Symbols CXCL5
Host Species Mouse
Immunogen Recombinant Human CXCL5 Protein.
Purification Method Protein A
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6374
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.