Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CXCL8/IL-8 Mouse anti-Bovine, Canine, Clone: MT8H6, Novus Biologicals™

Mouse Monoclonal Antibody
$335.76 - $1149.48
Specifications
| Antigen | CXCL8/IL-8 |
|---|---|
| Clone | MT8H6 |
| Concentration | 0.5 mg/ml |
| Applications | ELISA |
| Classification | Monoclonal |
Description
CXCL8/IL-8 Monoclonal antibody specifically detects CXCL8/IL-8 in Bovine, Canine samples. It is validated for ELISASpecifications
| CXCL8/IL-8 | |
| 0.5 mg/ml | |
| Monoclonal | |
| Purified | |
| RUO | |
| Bovine, Canine | |
| 3-10C, AMCF-I, C-X-C motif chemokine 8, CXCL8SCYB8, Emoctakin, GCP-1TSG-1, interleukin 8, K60, LECT, MDNCFb-ENAP, member 8, MONAPGCP1, NAP-1NAP1, Neutrophil-activating protein 1, Protein 3-10C, T cell chemotactic factor, T-cell chemotactic factor | |
| Recombinant human CXCL8/ IL-8 | |
| Primary | |
| Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles. |
| MT8H6 | |
| ELISA | |
| Unconjugated | |
| Mouse | |
| Angiogenesis, Biologically Active Proteins, Cancer, Chemokines and Cytokines, Cytokine Research, Immunology, Innate Immunity, Signal Transduction | |
| 0.2 um filtered PBS | |
| 3576 | |
| IgG1 | |
| Protein G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title