Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CXCL8/IL8 Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
$518.60
Specifications
Antigen | CXCL8/IL8 |
---|---|
Concentration | 0.4 mg/mL |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-5560
![]() ![]() |
Proteintech Group Inc
270951AP150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
Interleukin-8 (IL-8) is a chemokine produced by macrophages and other cell types such as epithelial cells. It is also synthesized by endothelial cells, which store IL-8 in their storage vesicles, the Weibel-Palade bodies. In humans, the interleukin-8 protein is encoded by the IL8 gene. Interleukin-8 (IL8) is a member of the CXC chemokine family. IL-8 is believed to play a role in the pathogenesis of bronchiolitis, a common respiratory tract disease caused by viral infection. This gene and other ten members of the CXC chemokine gene family form a chemokine gene cluster in a region mapped to chromosome 4q. The genes for IL8 have been co-localized on a 335-kb genomic fragment.Specifications
CXCL8/IL8 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
P10145 | |
3576 | |
CXCL8/IL8 Fusion Protein Ag25640 | |
Primary | |
-20°C | |
CXCL8 |
0.4 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
C X C motif chemokine 8, CXCL8, CXCL8,IL8, Emoctakin, GCP 1, GCP1, IL 8, IL 8(1 77), IL8, IL8/NAP1 form I Interleukin 8, IL8/NAP1 form II, IL8/NAP1 form VI, interleukin 8, MDNCF, MDNCF b IL 8(5 77) IL 8(6 77), NAF, NAF IL 8(7 77), NAP 1, NAP1, Neutrophil activating factor, Protein 3 10C | |
CXCL8 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title