Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CXCL9 Goat anti-Human, Polyclonal , Abnova™

Catalog No. 89934025 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-934-025 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-934-025 Supplier Abnova Corporation Supplier No. PAB29780

Goat polyclonal antibody raised against partial recombinant human CXCL9.

The function of this gene has not been specifically defined; however, it is thought to be involved in T cell trafficking. This gene has been localized to 4q21 with INP10, which is also a member of the chemokine family of cytokines. [provided by RefSeq]

Specifications

Antigen CXCL9
Applications ELISA
Classification Polyclonal
Conjugate Unconjugated
Formulation In PBS, pH 7.2 (0.09% sodium azide)
Gene Accession No. Q07325
Gene Alias CMK, Humig, MIG, SCYB9, crg10
Gene Symbols CXCL9
Host Species Goat
Immunogen Recombinant protein corresponding to partial human CXCL9.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4283
Target Species Human
Content And Storage Store at 4°C.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.