Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CXCL9/MIG Antibody (050), Alexa Fluor™ 750, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP289697AF750
Description
CXCL9/MIG Monoclonal specifically detects CXCL9/MIG in Human samples. It is validated for ELISA, Sandwich ELISA Detection.Specifications
CXCL9/MIG | |
Monoclonal | |
Alexa Fluor 750 | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Has cross-reactivity in ELISA with: Mouse CXCL9 Cynomolgus CXCL9 No cross-reactivity in ELISA with: Human CXCL10 | |
Store at 4°C in the dark. | |
IgG |
ELISA | |
50 | |
ELISA, Sandwich ELISA Detection | |
chemokine (C-X-C motif) ligand 9, CMK, crg-10, C-X-C motif chemokine 9, Gamma-interferon-induced monokine, Humig, MIGSmall-inducible cytokine B9, monokine induced by gamma interferon, SCYB9Monokine induced by interferon-gamma | |
This antibody was obtained from a rabbit immunized with purified, recombinant Human CXCL9/MIG (Q07325; Thr23-Thr125). | |
0.1 mL | |
Biologically Active Proteins, Chemokines and Cytokines, Signal Transduction | |
4283 | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction