Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CXCL9/MIG Antibody (277), Alexa Fluor™ 700, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290453AF700
Description
CXCL9/MIG Monoclonal specifically detects CXCL9/MIG in Mouse samples. It is validated for ELISA.Specifications
CXCL9/MIG | |
Monoclonal | |
Alexa Fluor 700 | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Has cross-reactivity in ELISA with: Human CXCL9 No cross-reactivity in ELISA with: Mouse CXCL2 Human CXCL2 | |
Store at 4°C in the dark. | |
IgG |
ELISA | |
277 | |
ELISA | |
chemokine (C-X-C motif) ligand 9, CMK, crg-10, C-X-C motif chemokine 9, Gamma-interferon-induced monokine, Humig, MIGSmall-inducible cytokine B9, monokine induced by gamma interferon, SCYB9Monokine induced by interferon-gamma | |
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse CXCL9/MIG (P18340; Thr22-Thr126). | |
0.1 mL | |
Biologically Active Proteins, Chemokines and Cytokines, Signal Transduction | |
4283 | |
Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction