Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CXCL9 (MIG) Monoclonal Antibody (C7), Invitrogen™

Mouse Monoclonal Antibody
$161.70 - $522.50
Specifications
| Antigen | CXCL9 (MIG) |
|---|---|
| Clone | C7 |
| Concentration | 1 mg/mL |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Monoclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
PIMA543864
|
Invitrogen™
MA543864 |
20 μL |
Each for $161.70
|
|
|||||
PIMA543863
![]() |
Invitrogen™
MA543863 |
100 μL |
Each for $522.50
|
|
|||||
Description
MIG, also known as CXCL9, is a T-cell chemoattractant inducible by gamma interferon that is a member of the CXC chemokine family of cytokines.This gene is mapped to 4q21. It is noted that, although the best-described activities of the chemokines are as chemotactic factors, chemokines also have an effect on T-cell activation, angiogenesis, and HIV infection. While most CXC chemokines are chemotactic for neutrophils, MIG and INP10 are unusual and similar in being CXC chemokines that are chemotactic for lymphocytes and inactive in neutrophils.Specifications
| CXCL9 (MIG) | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| BB139920; C Cmotif chemokine; C X C motif chemokine; CC motif chemokine; CCmotif chemokine; chemokine (C-X-C motif) ligand 9; CMK; crg-10; CXC; CXC motif chemokine; C-X-C motif chemokine 9; C-X-C motif chemokine ligand 9; CXCL; CXCL9; gamma interferon-induced monokine; gamma-interferon-induced monokine; HuMIG; M119; Mig; monokine induced by gamma interferon; monokine induced by interferon gamma; monokine induced by interferon-gamma; MuMIG; protein m119; SCYB9; small inducible cytokine B subfamily (Cys-X-Cys), member 9; small inducible cytokine B9; small-inducible cytokine B9 | |
| CXCL9 | |
| IgG2b κ | |
| Protein A/G | |
| Antibody |
| C7 | |
| Immunohistochemistry (Paraffin), Western Blot | |
| Unconjugated | |
| Mouse | |
| Human | |
| Q07325 | |
| 4283 | |
| Thr23-Thr125 (Accession Q07325). | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| CXCL9 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title