Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CXCL9 (MIG) Polyclonal Antibody
CXCL9 (MIG) Polyclonal Antibody
Supplier: Thermo Scientific PA579116
Description
CXCL9 (MIG) Polyclonal Antibody for Western Blot, ELISA

Specifications
CXCL9 (MIG) | |
Polyclonal | |
Unconjugated | |
CXCL9 | |
BB139920; C Cmotif chemokine; C X C motif chemokine; CC motif chemokine; CCmotif chemokine; chemokine (C-X-C motif) ligand 9; CMK; crg-10; CXC; CXC motif chemokine; C-X-C motif chemokine 9; C-X-C motif chemokine ligand 9; CXCL; CXCL9; gamma interferon-induced monokine; gamma-interferon-induced monokine; HuMIG; M119; Mig; monokine induced by gamma interferon; monokine induced by interferon gamma; monokine induced by interferon-gamma; MuMIG; protein m119; SCYB9; small inducible cytokine B subfamily (Cys-X-Cys), member 9; small inducible cytokine B9; small-inducible cytokine B9 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
4283 | |
-20°C | |
Lyophilized |
ELISA, Western Blot | |
500 μg/mL | |
PBS with 5mg BSA and 0.05mg sodium azide | |
Q07325 | |
CXCL9 | |
E. coli-derived human CXCL9 recombinant protein (Position: T23-T125). | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction