Learn More
Invitrogen™ CXCL9 Monoclonal Antibody (49106), FITC

Description
In ELISAs, does not cross-react with recombinant mouse (rm) CXCL9, recombinant human CXCL10.
Specifications
Specifications
| Antigen | CXCL9 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | 49106 |
| Conjugate | FITC |
| Formulation | Buffered saline solution with ≤0.5% BSA and 0.09% sodium azide |
| Gene | CXCL9 |
| Gene Accession No. | Q07325 |
| Gene Alias | BB139920; C Cmotif chemokine; C X C motif chemokine; CC motif chemokine; CCmotif chemokine; chemokine (C-X-C motif) ligand 9; CMK; crg-10; CXC; CXC motif chemokine; C-X-C motif chemokine 9; C-X-C motif chemokine ligand 9; CXCL; CXCL9; gamma interferon-induced monokine; gamma-interferon-induced monokine; HuMIG; M119; Mig; monokine induced by gamma interferon; monokine induced by interferon gamma; monokine induced by interferon-gamma; MuMIG; protein m119; SCYB9; small inducible cytokine B subfamily (Cys-X-Cys), member 9; small inducible cytokine B9; small-inducible cytokine B9 |
| Gene Symbols | CXCL9 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.