Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CXCR-4, Rabbit anti-Human, Polyclonal, Millipore Sigma™

Catalog No. ABN144925UL Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABN144925UL 25 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABN144925UL Supplier MilliporeSigma Supplier No. ABN144925UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-CXCR-4 Antibody, Cat. No. ABN1449, is a highly specific rabbit polyclonal antibody that targets human CXCR-4 and has been tested in Immunohistochemistry (Paraffin) and Western Blotting.

C-X-C chemokine receptor type 4 (UniProt: P61073; also known as CXC-R4, CXCR-4, FB22, Fusin, HM89, LCR1, Leukocyte-derived seven transmembrane domain receptor, LESTR, Lipopolysaccharide-associated protein 3, LAP-3, LPS-associated protein 3, NPYRL, Stromal cell-derived factor 1 receptor, SDF-1 receptor, CD184) is encoded by the CXCR4 gene (Gene ID: 7852) in human. CXCR-4 is a receptor for the C-X-C chemokine CXCL12/SDF-1 that transduces a signal by increasing intracellular calcium levels and enhancing MAPK1/MAPK3 activation. CXCR-4 is expressed in numerous tissues and two isoforms have been reported that are generated by alternative splicing. Isoform 1 is the predominant form in all tissues tested. CXCR-4 is rapidly phosphorylated on serine and threonine residues in the C-terminal upon agonist stimulation. Phosphorylation at Ser324 and Ser325 leads to recruitment of ITCH (E3 ubiquitin-protein ligase), ubiquitination and protein degradation. Sulfation on Tyr21 is shown to be essential for efficient binding of CXCL12/SDF-1alpha and it promotes its dimerization. Tyr7 and Tyr12 are sulfated in a sequential manner following complete sulfation of Tyr21. CXCR-4 acts as a co-receptor for human HIV-1 virus isolates (CD4 being the primary receptor) and as a primary receptor for HIV-2 isolates. CXCR-4 can also bind bacterial lipopolysaccharide (LPS) and mediate LPS-induced inflammatory response, including TNF-alpha secretion by monocytes. Mutations in CXCR4 gene cause WHIM syndrome that is characterized by neutropenia, hypogammaglobulinemia and extensive human papillomavirus (HPV) infection.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CXCR-4
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Immunohistochemistry (Paraffin) Analysis: A 1:250 dilution from a representative lot detected CXCR-4 in human bone marrow, human pancreas, human breast and human prostate tissues.
Formulation Purified rabbit polyclonal antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.
Gene Alias C-X-C chemokine receptor type 4;CXC-R4;FB22;Fusin;HM89;LCR1;Leukocyte-derived seven transmembrane domain receptor;LESTR;Lipopolysaccharide-associated protein 3;LAP-3;LPS-associated protein 3;NPYRL;Stromal cell-derived factor 1 receptor;SDF-1 receptor;CD184
Gene Symbols CXCR4
Host Species Rabbit
Immunogen KLH-conjugated linear peptide corresponding to 12 amino acids from the N-terminal region of human CXCR-4.
Molecular Weight of Antigen ∽53 kDa observed; 39.75 kDa calculated. Calculated molecular weight is 40 kDa but we observed it larger at around ∽50 kDa.Uncharacterized bands may be observed in some lysate(s).
Purification Method Affinity Purified
Quantity 25 μL
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Test Specificity This rabbit polyclonal antibody detects CXCR-4 in human cells. It targets an epitope within 12 amino acids from the N-terminal region. Immunogen sequence is conserved between isoforms 1 and 2. Hence, it may react with both isoforms.
Target Species Human
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.