Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CXCR1/IL-8RA Antibody (8F1-1-4) - BSA Free, Novus Biologicals™
SDP

Mouse Monoclonal Antibody

$206.50 - $499.50

Specifications

Antigen CXCR1/IL-8 RA
Clone 8F1-1-4
Concentration 0.5 mg/mL
Dilution Flow Cytometry 1:10-1:1000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500, Fluorophore-linked immunosorbent assay 1:100-1:2000
Applications Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunofluorescence
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP14333903
SDP
View Documents
Novus Biologicals
NBP1433390.025MG
0.025 mg
Each for $206.50
Only null left
Add to Cart
 
NBP14333901
SDP
View Documents
Novus Biologicals
NBP1433390.1MG
0.1 mg
Each for $499.50
Only null left
Add to Cart
 
Description

Description

CXCR1/IL-8RA Monoclonal specifically detects CXCR1/IL-8RA in Human, Hamster samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin, Fluorophore-linked immunosorbent assay.
Specifications

Specifications

CXCR1/IL-8 RA
0.5 mg/mL
Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunofluorescence
Unconjugated
RUO
Human
P25024
3577
The immunogen for this antibody was CD181.
Primary
Store at 4C. Do not freeze.
8F1-1-4
Flow Cytometry 1:10-1:1000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500, Fluorophore-linked immunosorbent assay 1:100-1:2000
Monoclonal
Mouse
Cytokine Research, GPCR, Immunology, Innate Immunity, Signal Transduction
Aqueous, may contain carrier protein/stabilizer. with 0.09% Sodium Azide
CD128, CD181, CD181 antigen, CDw128aC-C, chemokine (C-X-C motif) receptor 1, CKR-1, CMKAR1, C-X-C chemokine receptor type 1, CXC-R1, CXCR-1, High affinity interleukin-8 receptor A, IL-8 receptor type 1, IL-8R A, IL8R1, IL8RAC-C-CKR-1, IL8RBA, interleukin 8 receptor, alpha, interleukin-8 receptor type 1, interleukin-8 receptor type A
CXCR1
IgG2b κ
Affinity Purified
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.