Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CXCR1/IL-8RA Antibody (SE2), DyLight 650, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP312003C
Description
CXCR1/IL-8RA Monoclonal antibody specifically detects CXCR1/IL-8RA in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, ImmunofluorescenceSpecifications
CXCR1/IL-8RA | |
Monoclonal | |
DyLight 650 | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human | |
IgG κ |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence | |
SE2 | |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
CD128, CD181, CD181 antigen, CDw128aC-C, chemokine (C-X-C motif) receptor 1, CKR-1, CMKAR1, C-X-C chemokine receptor type 1, CXC-R1, CXCR-1, High affinity interleukin-8 receptor A, IL-8 receptor type 1, IL-8R A, IL8R1, IL8RAC-C-CKR-1, IL8RBA, interleukin 8 receptor, alpha, interleukin-8 receptor type 1, interleukin-8 receptor type A | |
This antibody was raised by immunizing BALB/c mice with a BSA-conjugated peptide comprising of the N-terminal part of CXCR1/IL-8RA (amino acids 1-30). | |
0.1 mL | |
Chemokines and Cytokines, Cytokine Research, GPCR, Immunology, Innate Immunity, Signal Transduction | |
3577 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction