Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CXCR1/IL-8RA Mouse anti-Human, Clone: SE2, Novus Biologicals™

Mouse Monoclonal Antibody
$268.00 - $584.50
Specifications
Antigen | CXCR1/IL-8RA |
---|---|
Clone | SE2 |
Concentration | 1 mg/ml |
Dilution | Western Blot, Flow Cytometry, Immunohistochemistry |
Applications | Western Blot, Flow Cytometry, Immunohistochemistry |
Description
CXCR1/IL-8RA Monoclonal antibody specifically detects CXCR1/IL-8RA in Human samples. It is validated for Western Blot, Flow Cytometry, ImmunohistochemistrySpecifications
CXCR1/IL-8RA | |
1 mg/ml | |
Western Blot, Flow Cytometry, Immunohistochemistry | |
Unconjugated | |
Mouse | |
Chemokines and Cytokines, Cytokine Research, GPCR, Immunology, Innate Immunity, Signal Transduction | |
PBS | |
3577 | |
IgG2b κ | |
Protein A purified |
SE2 | |
Western Blot, Flow Cytometry, Immunohistochemistry | |
Monoclonal | |
Purified | |
RUO | |
Human | |
CD128, CD181, CD181 antigen, CDw128aC-C, chemokine (C-X-C motif) receptor 1, CKR-1, CMKAR1, C-X-C chemokine receptor type 1, CXC-R1, CXCR-1, High affinity interleukin-8 receptor A, IL-8 receptor type 1, IL-8R A, IL8R1, IL8RAC-C-CKR-1, IL8RBA, interleukin 8 receptor, alpha, interleukin-8 receptor type 1, interleukin-8 receptor type A | |
This antibody was raised by immunizing BALB/c mice with a BSA-conjugated peptide comprising of the N-terminal part of CXCR1/IL-8RA (amino acids 1-30). | |
Primary | |
Store at 4C for up to 3 months. For longer storage, aliquot and store at -20C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title