Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CXCR1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5144682
Description
Positive control: U87-MG cell lysate, Hela cell lysate, Jurkat cell lysate, HepG2 cell lysate, Hela. Subcellular Location: Cell membrane.
CXCR1 is a member of the G-protein-coupled receptor family. This protein is a receptor for interleukin 8 (IL8). It binds to IL8 with high affinity, and transduces the signal through a G-protein activated second messenger system. Knockout studies in mice suggested that this protein inhibits embryonic oligodendrocyte precursor migration in developing spinal cord. This gene, IL8RB, a gene encoding another high affinity IL8 receptor, as well as IL8RBP, a pseudogene of IL8RB, form a gene cluster in a region mapped to chromosome 2q33-q36.
Specifications
CXCR1 | |
Polyclonal | |
Unconjugated | |
CXCR1 | |
C Cmotif chemokine; C X C motif chemokine; C-C; CC motif chemokine; C-C-CKR-1; CCmotif chemokine; CD128; CD181; CDw128a; chemokine (C-X-C motif) receptor 1; CKR-1; CMKAR1; CXC; CXC chemokine receptor 1; C-X-C chemokine receptor type 1; CXC motif chemokine; C-X-C motif chemokine receptor 1; CXCR1; CXC-R1; CXCR-1; High affinity interleukin-8 receptor A; IL-8 receptor; IL-8 Receptor alpha; IL-8 Receptor IL-RA; IL-8 receptor type 1; IL-8R A; IL8R1; Il8ra; IL8RBA; IL-RA; interleukin 8 receptor; interleukin 8 receptor alpha; interleukin 8 receptor, alpha; interleukin-8 receptor type 1; interleukin-8 receptor type A | |
Rabbit | |
Affinity Chromatography | |
RUO | |
3577 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot, Immunocytochemistry | |
2 mg/mL | |
PBS with 0.1% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4 | |
P25024 | |
CXCR1 | |
Recombinant protein within full length human CXCR1 protein aa 1-350/350. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction