Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CXCR2/IL-8RB Antibody (48311.211), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB1001631
Description
CXCR2/IL-8RB Monoclonal specifically detects CXCR2/IL-8RB in Human samples. It is validated for Flow Cytometry, Block/Neutralize.Specifications
CXCR2/IL-8RB | |
Monoclonal | |
LYOPH | |
Flow Cytometry 1:10-1:1000, Block/Neutralize 0.5 - 1.5 ul/ml | |
CD182 antigen, CDw128b, chemokine (C-X-C motif) receptor 2, chemokine (CXC) receptor 2, C-X-C chemokine receptor type 2, CXC-R2, CXCR-2, CXCR2 gene for IL8 receptor type B, GRO/MGSA receptor, High affinity interleukin-8 receptor B, IL-8 receptor type 2, IL-8R B, IL8R2, IL8RA, interleukin 8 receptor B, interleukin 8 receptor type 2, interleukin-8 receptor type B | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
Monoclonal Anti-Human CXCR-2 reacts with CXCR-2 transfected cells and not with the parent cell line by flow cytometry. The antibody shows no cross-reactivity with human CXCR-1. | |
Human | |
Purified |
Flow Cytometry, Blocking Assay | |
48311.211 | |
Unconjugated | |
Lyophilized from 0.2 mm-filtered solution in phosphate buffered saline containing carbohydrates. with 0.05% Sodium Azide | |
CXCR2 | |
Human CXCR2 transfected NSO mouse myeloma cells. | |
0.1 mg | |
Cancer, Chemokines and Cytokines, Cytokine Research, GPCR, Immunology, Innate Immunity | |
3579 | |
Reconstitute with 1.0 ml of 0.2 um-filtered PBS to a final concentration of 0.5 mg/ml. | |
Store at -20C. Avoid freeze-thaw cycles. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction