Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CXCR2/IL-8RB Antibody (866614) [DyLight 405], Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB8110E
Description
CXCR2/IL-8RB Monoclonal antibody specifically detects CXCR2/IL-8RB in Rat samples. It is validated for Flow Cytometry, CyTOFSpecifications
CXCR2/IL-8RB | |
Monoclonal | |
DyLight 405 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Primary | |
Rat | |
Purified |
Flow Cytometry, CyTOF | |
866614 | |
Flow Cytometry, CyTOF-ready | |
CD182 antigen, CDw128b, chemokine (C-X-C motif) receptor 2, chemokine (CXC) receptor 2, C-X-C chemokine receptor type 2, CXC-R2, CXCR-2, CXCR2 gene for IL8 receptor type B, GRO/MGSA receptor, High affinity interleukin-8 receptor B, IL-8 receptor type 2, IL-8R B, IL8R2, IL8RA, interleukin 8 receptor B, interleukin 8 receptor type 2, interleukin-8 receptor type B | |
NS0 mouse myeloma cell line transfected with rat CXCR2/IL-8 RB, Accession # P35407 | |
0.1 mL | |
Angiogenesis, Cancer, Chemokines and Cytokines, Cytokine Research, GPCR, Immunology, Innate Immunity, Myeloid derived Suppressor Cell | |
3579 | |
Store at 4°C in the dark. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction