Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CXCR2/IL-8RB Rabbit anti-Human, Clone: HC2, Novus Biologicals™

Rabbit Monoclonal Antibody
$269.00 - $584.50
Specifications
Antigen | CXCR2/IL-8RB |
---|---|
Clone | HC2 |
Dilution | Flow Cytometry, Immunocytochemistry/Immunofluorescence |
Classification | Monoclonal |
Conjugate | Unconjugated |
Description
CXCR2/IL-8RB Monoclonal antibody specifically detects CXCR2/IL-8RB in Human samples. It is validated for Flow Cytometry, ImmunofluorescenceSpecifications
CXCR2/IL-8RB | |
Flow Cytometry, Immunocytochemistry/Immunofluorescence | |
Unconjugated | |
RUO | |
Human | |
CD182 antigen, CDw128b, chemokine (C-X-C motif) receptor 2, chemokine (CXC) receptor 2, C-X-C chemokine receptor type 2, CXC-R2, CXCR-2, CXCR2 gene for IL8 receptor type B, GRO/MGSA receptor, High affinity interleukin-8 receptor B, IL-8 receptor type 2, IL-8R B, IL8R2, IL8RA, interleukin 8 receptor B, interleukin 8 receptor type 2, interleukin-8 receptor type B | |
This antibody was raised by immunizing mice with human IL-8 (interleukin-8) receptor, type B. | |
Primary |
HC2 | |
Monoclonal | |
Rabbit | |
Angiogenesis, Cancer, Chemokines and Cytokines, Cytokine Research, GPCR, Immunology, Innate Immunity, Myeloid derived Suppressor Cell | |
PBS | |
3579 | |
IgG κ | |
Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title