Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CXCR2 Monoclonal Antibody (C8), Invitrogen™
Mouse Monoclonal Antibody
$195.80 - $437.00
Specifications
Antigen | CXCR2 |
---|---|
Clone | C8 |
Concentration | 1 mg/mL |
Applications | Western Blot, Immunohistochemistry (Paraffin), Western Blot |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
FEMA551156
|
Invitrogen™
MA551156 |
20 μL |
Each for $195.80
|
|
|||||
FEMA551155
|
Invitrogen™
MA551155 |
200 μL |
Each for $437.00
|
|
|||||
Description
CXCR2 is a receptor for interleukin-8, which is a powerful neutrophil chemotactic factor. It is a member of the GPCR family. Binding of IL-8 to the receptor causes activation of neutrophils. This response is mediated via a G-protein that activate a phosphatidylinositol-calcium second messenger system. This receptor binds to IL-8 with a high affinity and to GRO/MGSA and NAP-2 also with a high affinity. It has been reported to be expressed in a wide variety of tissues. ESTs have been isolated from human placenta and thymus libraries.Specifications
CXCR2 | |
1 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.05% Proclin 300; pH 7.4 | |
C Cmotif chemokine; C X C motif chemokine; CC motif chemokine; CCmotif chemokine; CD128; CD182; CD182 antigen; CDw128; CDw128b; chemokine (C-X-C motif) receptor 2; chemokine (CXC) receptor 2; chemokine (C-X-C) receptor 2; Cmkar2; CXC; C-X-C chemokine receptor type 2; CXC motif chemokine; C-X-C motif chemokine receptor 2; Cxcr2; CXC-R2; CXCR-2; CXCR2 gene for IL8 receptor type B; Gpcr16; GRO/MGSA receptor; high affinity interleukin-8 receptor B; IL-8 receptor alpha chain; Il-8 receptor b; IL-8 receptor beta; IL-8 receptor type 2; IL-8R B; IL8R2; IL8RA; Il8rb; IL-8rb; IL-8Rh; interleukin 8 receptor B; interleukin 8 receptor type 2; interleukin 8 receptor, beta; interleukin-8 receptor type B; mIL-8RH | |
CXCR2 | |
IgG1 κ | |
Protein A/G | |
CXCR2 |
C8 | |
Western Blot, Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Mouse | |
Human, Pig | |
P25025 | |
100124654, 3579 | |
Recombinant Human IL8Rb protein, Met1-Leu57 (Accession #P25025). | |
Primary | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title