Learn More
CXCR3 (CD183) Armenian Hamster anti-Mouse, Unconjugated, Clone: 173, MilliporeSigma™
Shop All MilliporeSigma ProductsDescription
Specifications
Specifications
| Antigen | CXCR3 (CD183) |
| Applications | Flow Cytometry, Inhibition Assays, Radioimmune Assays (RIA) |
| Classification | Monoclonal |
| Clone | 173 |
| Conjugate | Unconjugated |
| Gene Accession No. | NP_034040 |
| Gene Alias | C-X-C chemokine receptor type 3;CXC-R3;Interferon-inducible protein 10 receptor;IP-10 receptor;CD183 |
| Gene Symbols | Cxcr3, Cmkar3 |
| Host Species | Armenian Hamster |
| Immunogen | A multiple antigen peptide (MAPS8) corresponding to 37 amino acids from the N-terminal region of murine C-X-C chemokine receptor 3 (CXCR3). |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.