Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CXCR3 (CD183) Armenian Hamster anti-Mouse, Unconjugated, Clone: 173, MilliporeSigma™

Catalog No. MABF21812MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABF21812MI 25 μL
MABF21811MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MABF21812MI Supplier MilliporeSigma Supplier No. MABF218125UL
Add to Cart
Add to Cart

Armenian Hamster Monoclonal Antibody

CXCR3 (CD183) Monoclonal antibody specifically detects CXCR3 (CD183) in Mouse samples. It is validated for Flow Cytometry, Inhibition, Radioimmunoassay.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CXCR3 (CD183)
Applications Flow Cytometry, Inhibition Assays, Radioimmune Assays (RIA)
Classification Monoclonal
Clone 173
Conjugate Unconjugated
Gene Accession No. NP_034040
Gene Alias C-X-C chemokine receptor type 3;CXC-R3;Interferon-inducible protein 10 receptor;IP-10 receptor;CD183
Gene Symbols Cxcr3, Cmkar3
Host Species Armenian Hamster
Immunogen A multiple antigen peptide (MAPS8) corresponding to 37 amino acids from the N-terminal region of murine C-X-C chemokine receptor 3 (CXCR3).
Molecular Weight of Antigen 41.02 kDa calculated.
Purification Method Protein G Purified
Quantity 25 μL
Research Discipline Inflammation and Immunology
Primary or Secondary Primary
Test Specificity Clone 173 is an Armenian Hamster monoclonal antibody that specifically detects CXCR3 on cells from wild-type, but not from CXCR3-/- mice.
Target Species Mouse
Content And Storage Stable for 1 year at -20°C from date of receipt.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.