Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CXCR3 Mouse anti-Human, Clone:5C10E6, Abnova™
Mouse monoclonal antibody raised against recombinant human CXCR3.
Supplier: Abnova Corporation MAB17971
Description
This gene encodes a G protein-coupled receptor with selectivity for three chemokines, termed IP10 (interferon-g-inducible 10 kDa protein), Mig (monokine induced by interferon-g) and I-TAC (interferon-inducible T cell a-chemoattractant). IP10, Mig and I-TAC belong to the structural subfamily of CXC chemokines, in which a single amino acid residue separates the first two of four highly conserved Cys residues. Binding of chemokines to this protein induces cellular responses that are involved in leukocyte traffic, most notably integrin activation, cytoskeletal changes and chemotactic migration. Inhibition by Bordetella pertussis toxin suggests that heterotrimeric G protein of the Gi-subclass couple to this protein. Signal transduction has not been further analyzed but may include the same enzymes that were identified in the signaling cascade induced by other chemokine receptors. As a consequence of chemokine-induced cellular desensitization (phosphorylation-dependent receptor internalization), cellular responses are typically rapid and short in duration. Cellular responsiveness is restored after dephosphorylation of intracellular receptors and subsequent recycling to the cell surface. This gene is prominently expressed in in vitro cultured effector/memory T cells, and in T cells present in many types of inflamed tissues. In addition, IP10, Mig and I-TAC are commonly produced by local cells in inflammatory lesion, suggesting that this gene and its chemokines participate in the recruitment of inflammatory cells. Therefore, this protein is a target for the development of small molecular weight antagonists, which may be used in the treatment of diverse inflammatory diseases. Multiple transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq]Specifications
CXCR3 | |
Monoclonal | |
Unconjugated | |
ELISA (1:10000) Flow Cytometry (1:200-1:400) Western Blot (1:100-1:500) The optimal working dilution should be determined by the end user. |
|
CXCR3 | |
Mouse | |
100 μg | |
Human | |
Antibody | |
IgG1 |
ELISA, Flow Cytometry, Western Blot | |
5C10E6 | |
chemokine (C-X-C motif) receptor 3 | |
In PBS (0.05% sodium azide) | |
CD182, CD183, CKR-L2, CMKAR3, GPR9, IP10-R, Mig-R, MigR | |
Recombinant protein corresponding to human CXCR3. | |
2833 | |
Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction