Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Rat CXCR3 Alexa Fluor 700-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems FAB8109N100UG
This item is not returnable.
View return policy
Description
CXCR3 Monoclonal specifically detects CXCR3 in Rat samples. It is validated for Flow Cytometry.Specifications
CXCR3 | |
Monoclonal | |
0.2 mg/mL | |
Flow Cytometry 0.25-1 ug/10^6 cells | |
Q9J119 | |
CXCR3 | |
NS0 mouse myeloma cell line transfected with rat CXCR3 Accession # Q9J119 | |
RUO | |
2833 | |
Rat | |
IgG2b |
Flow Cytometry | |
868013 | |
Alexa Fluor 700 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
CD183, CD183 antigen, chemokine (C-X-C motif) receptor 3, chemokine (C-X-C) receptor 3, CKR-L2IP-10 receptor, CMKAR3, C-X-C chemokine receptor type 3, CXCR3, CXC-R3, CXCR-3, G protein-coupled receptor 9CD182, GPR9, GPR9Mig-R, Interferon-inducible protein 10 receptor, IP10 receptor, IP10-R, Mig receptor, MigR | |
Mouse | |
100 μg | |
Primary | |
Detects rat CXCR3 in ELISA. Stains rat CXCR3 transfected cells but not irrelevant transfectants by Flow Cytometry. | |
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction