Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CXCR3 Polyclonal Antibody
CXCR3 Polyclonal Antibody
Supplier: Thermo Scientific PA595185
Description
CXCR3 Polyclonal Antibody for Western Blot, IHC (P)

Specifications
CXCR3 | |
Polyclonal | |
Unconjugated | |
CXCR3 | |
an; C Cmotif chemokine; C X C motif chemokine; CC motif chemokine; CCmotif chemokine; CD182; CD183; chemokine (C-X-C motif) receptor 3; chemokine receptor 3; CKR-L2; Cmkar3; CXC; C-X-C chemokine receptor type 3; CXC motif chemokine; C-X-C motif chemokine receptor 3; Cxcr3; CXC-R3; CXCR-3; G protein-coupled receptor 9; GPR9; Interferon-inducible protein 10 receptor; IP10; IP10 receptor; IP-10 receptor; IP10-R; Mig receptor; MigR; Mig-R | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
2833, 84475 | |
-20°C | |
Lyophilized |
Immunohistochemistry (Paraffin), Western Blot | |
500 μg/mL | |
PBS with 5mg BSA and 0.05mg sodium azide | |
P49682, Q9JII9 | |
CXCR3 | |
E.coli-derived human CXCR3 recombinant protein (Position: M1-L368). | |
100 μg | |
Primary | |
Human, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction