Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CXCR3 Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
$518.60
Specifications
Antigen | CXCR3 |
---|---|
Concentration | 0.4 mg/mL |
Applications | Immunohistochemistry (Paraffin), Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-5445
![]() ![]() |
Proteintech Group Inc
267561AP150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
Chemokines play important roles in inflammation and critical for the recruitment of effector immune cells to sites of infection. Historically, CD183 was the third CXC chemokine receptor discovered and therefore, commonly designated as CXCR3. Binding of chemokines to CD183 induces cellular responses that are involved in leukocyte traffic, most notably integrin activation, cytoskeletal changes and chemotactic migration. The CXC chemokine receptor (CXCR) 3, a G-protein-coupled 7-transmembrane receptor, is expressed at high levels on activated and memory T cells, B cells, natural killer cells, and plasmacytoid monocytes.Specifications
CXCR3 | |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse | |
O88410, P49682 | |
12766, 2833 | |
CXCR3 Fusion Protein Ag25056 | |
Primary | |
-20°C | |
CXCR3 |
0.4 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
CD183, CKR L2, CMKAR3, CXC R3, CXCR 3, CXCR3, CXCR3A, CXCR3B, G protein coupled receptor 9, GPR9, IP 10 receptor, IP10 R, Mig R, MigR | |
CXCR3 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title