Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CXCR3 Rabbit anti-Human, Mouse, Rat, Clone: 3B7Y7, Novus Biologicals™

Rabbit Monoclonal Antibody
$220.54 - $523.50
Specifications
| Antigen | CXCR3 |
|---|---|
| Clone | 3B7Y7 |
| Dilution | Western Blot 1:500 - 1:2000, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:50 - 1:200 |
| Applications | Western Blot, Immunohistochemistry, Immunofluorescence |
| Classification | Monoclonal |
Description
CXCR3 Monoclonal antibody specifically detects CXCR3 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, ImmunofluorescenceSpecifications
| CXCR3 | |
| Western Blot 1:500 - 1:2000, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:50 - 1:200 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| CD183, CD183 antigen, chemokine (C-X-C motif) receptor 3, chemokine (C-X-C) receptor 3, CKR-L2IP-10 receptor, CMKAR3, C-X-C chemokine receptor type 3, CXC-R3, CXCR-3, G protein-coupled receptor 9CD182, GPR9Mig-R, Interferon-inducible protein 10 receptor, IP10 receptor, IP10-R, Mig receptor, MigR | |
| A synthetic peptide corresponding to a sequence within amino acids 200-300 of human CXCR3 (P49682). ATHCQYNFPQVGRTALRVLQLVAGFLLPLLVMAYCYAHILAVLLVSRGQRRLRAMRLVVVVVVAFALCWTPYHLVVLVDILMDLGALARNCGRESRVDVAK | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| 3B7Y7 | |
| Western Blot, Immunohistochemistry, Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Chemokines and Cytokines, Cytokine Research, GPCR, Immunology, Innate Immunity | |
| PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
| 2833 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title