Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CXCR4 Antibody (247506R), PE/Cy7, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals FAB21651RPECY7
Description
CXCR4 Monoclonal antibody specifically detects CXCR4 in Mouse samples. It is validated for Flow CytometrySpecifications
CXCR4 | |
Monoclonal | |
PE-Cyanine7 | |
PBS | |
Rat | |
Protein A or G purified from cell culture supernatant | |
RUO | |
Primary | |
Mouse | |
Purified |
Flow Cytometry | |
247506R | |
Flow Cytometry | |
CD184, chemokine (C-X-C motif) receptor 4, chemokine (C-X-C motif), receptor 4 (fusin), C-X-C chemokine receptor type 4, CXC-R4, CXCR-4, D2S201ESDF-1 receptor, FB22, Fusin, HM89NPYRL, HSY3RR, LAP3, LCR1, LESTRCD184 antigen, Leukocyte-derived seven transmembrane domain receptor, leukocyte-derived seven-transmembrane-domain receptor, lipopolysaccharide-associated protein 3, neuropeptide Y receptor Y3, NPY3RWHIM, NPYR, NPYY3R, pCXCR4, seven transmembrane helix receptor, seven-transmembrane-segment receptor, spleen, Stromal cell-derived factor 1 receptor | |
Y3 rat myeloid cell line transfected with mouse CXCR4, Met1-Ser359, Accession # P70658 | |
0.1 mL | |
Adaptive Immunity, Cancer, Chemokines and Cytokines, Cytokine Research, GPCR, Growth and Development, Immunology, Innate Immunity, Metastasis, Neuronal Cell Markers, Signal Transduction, Stem Cells | |
7852 | |
Store at 4C in the dark. Do not freeze. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction