Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CXCR4 Antibody

Goat Polyclonal Antibody has been used in 3 publications
$158.00
Specifications
Antigen | CXCR4 |
---|---|
Concentration | 1.0 mg/mL |
Dilution | Western Blot 1:250, ELISA 1:25000, Immunohistochemistry 1:150, Immunohistochemistry-Paraffin 1:150 |
Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), ELISA |
Classification | Polyclonal |
Description
CXCR4 Polyclonal specifically detects CXCR4 in Human, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
CXCR4 | |
Western Blot 1:250, ELISA 1:25000, Immunohistochemistry 1:150, Immunohistochemistry-Paraffin 1:150 | |
Polyclonal | |
Goat | |
Adaptive Immunity, Cancer, Cytokine Research, GPCR, Growth and Development, Immunology, Innate Immunity, Neuronal Cell Markers, Signal Transduction, Stem Cells | |
10mM KHPO4, 0.14M NaCl and 1.0 mg/ml BSA with 0.1% Sodium Azide | |
7852 | |
Synthetic peptide (TEEMGSGDYDSMKEPCFREENANFNKC) corresponding to N-terminal extracellular domain of human CXCR4 receptor. | |
Primary | |
Store at -20C. Avoid freeze-thaw cycles. |
1.0 mg/mL | |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), ELISA | |
Unconjugated | |
RUO | |
Human, Rat | |
P61073 | |
CXCR4 | |
IgG | |
Affinity Purified | |
Peptide sequence is < 50% identical to other human chemokine receptors in this region |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title