Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CXCR4 Antibody, Alexa Fluor™ 532, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP177560AF532
Description
CXCR4 Polyclonal specifically detects CXCR4 in Human samples. It is validated for Western Blot.Specifications
CXCR4 | |
Polyclonal | |
Western Blot | |
CD184, chemokine (C-X-C motif) receptor 4, chemokine (C-X-C motif), receptor 4 (fusin), C-X-C chemokine receptor type 4, CXC-R4, CXCR-4, D2S201ESDF-1 receptor, FB22, Fusin, HM89NPYRL, HSY3RR, LAP3, LCR1, LESTRCD184 antigen, Leukocyte-derived seven transmembrane domain receptor, leukocyte-derived seven-transmembrane-domain receptor, lipopolysaccharide-associated protein 3, neuropeptide Y receptor Y3, NPY3RWHIM, NPYR, NPYY3R, pCXCR4, seven transmembrane helix receptor, seven-transmembrane-segment receptor, spleen, Stromal cell-derived factor 1 receptor | |
Rabbit | |
0.1 mL | |
7852 | |
Store at 4°C in the dark. |
Western Blot | |
Alexa Fluor 532 | |
50 mM sodium borate with 0.05% sodium azide | |
CXCR4 | |
A synthetic peptide made to a C-terminal portion of the human CXCR4 protein (between residues 300-352) [UniProtNo. P61073] | |
Primary | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction