Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CXCR4 Antibody, Biotin, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP177560B
Description
CXCR4 Polyclonal specifically detects CXCR4 in Human samples. It is validated for Western Blot, Simple Western.Specifications
CXCR4 | |
Polyclonal | |
Western Blot, Simple Western | |
P61073 | |
CXCR4 | |
A synthetic peptide made to a C-terminal portion of the human CXCR4 protein (between residues 300-352) [UniProt# P61073] | |
0.1 mL | |
Adaptive Immunity, Cancer, Cytokine Research, GPCR, Growth and Development, Immunology, Innate Immunity, Neuronal Cell Markers, Signal Transduction, Stem Cells | |
7852 | |
Store at 4C in the dark. |
Western Blot | |
Biotin | |
PBS with 0.05% Sodium Azide | |
CD184, chemokine (C-X-C motif) receptor 4, chemokine (C-X-C motif), receptor 4 (fusin), C-X-C chemokine receptor type 4, CXC-R4, CXCR-4, D2S201ESDF-1 receptor, FB22, Fusin, HM89NPYRL, HSY3RR, LAP3, LCR1, LESTRCD184 antigen, Leukocyte-derived seven transmembrane domain receptor, leukocyte-derived seven-transmembrane-domain receptor, lipopolysaccharide-associated protein 3, neuropeptide Y receptor Y3, NPY3RWHIM, NPYR, NPYY3R, pCXCR4, seven transmembrane helix receptor, seven-transmembrane-segment receptor, spleen, Stromal cell-derived factor 1 receptor | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction