Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CXCR4 Goat anti-Human, Mouse, Polyclonal, Novus Biologicals™

Goat Polyclonal Antibody
$220.00
Specifications
Antigen | CXCR4 |
---|---|
Concentration | 1.0 mg/mL |
Dilution | Western Blot 1:500, ELISA 1:25000, Immunohistochemistry 1:150, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunohistochemistry-Paraffin 1:150 |
Applications | ELISA, Immunocytochemistry, Immunohistochemistry, Immunofluorescence, Western Blot, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Description
CXCR4 Polyclonal specifically detects CXCR4 in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
CXCR4 | |
Western Blot 1:500, ELISA 1:25000, Immunohistochemistry 1:150, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunohistochemistry-Paraffin 1:150 | |
Polyclonal | |
Concentrated | |
RUO | |
Human, Mouse | |
P70658 | |
CXCR4 | |
Primary | |
This binds to CXCR 4 receptor on mouse spleen leukocytes as determined by immunocytochemistry. |
1.0 mg/mL | |
ELISA, Immunocytochemistry, Immunohistochemistry, Immunofluorescence, Western Blot, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Goat | |
Adaptive Immunity, Cancer, Cytokine Research, GPCR, Growth and Development, Immunology, Innate Immunity, Neuronal Cell Markers, Signal Transduction, Stem Cells | |
10mM KHPO4, 0.14M NaCl and 1.0 mg/ml BSA with 0.1% Sodium Azide | |
7852 | |
IgG | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title