Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CXCR4 Mouse anti-Human, Clone: 4B5E4, Proteintech
SDP

Catalog No. 501736201 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-6201 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-6201 Supplier Proteintech Group Inc Supplier No. 600421IG150UL
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CXCR4 may be involved in WHIM syndrome (WHIMS). Immunodeficiency disease characterized by neutropenia, hypogammaglobulinemia and extensive human papillomavirus (HPV) infection. Despite the peripheral neutropenia, bone marrow aspirates from affected individuals contain abundant mature myeloid cells, a condition termed myelokathexis. The disease is caused by mutations affecting the gene represented in this entry.

Specifications

Antigen CXCR4
Applications Flow Cytometry, Western Blot
Classification Monoclonal
Clone 4B5E4
Concentration 1.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene CXCR4
Gene Accession No. P61073
Gene Alias CD184, CXC R4, CXCR 4, CXCR4, CXCR4B, D2S201E, FB22, Fusin, HM89, HSY3RR, LAP3, LCR1, LESTR, SDF 1 receptor, WHIM
Gene Symbols CXCR4
Host Species Mouse
Immunogen CXCR4 Fusion Protein Ag1528
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7852
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgM
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.