Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CXCR5 (CD185) Rat anti-Human, BB700, Clone: RF8B2, BD Horizon™

Catalog No. BDB566469 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 Tests
25 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB566469 100 Tests
BDB566470 25 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB566469 Supplier BD Biosciences Supplier No. 566469
Add to Cart
Add to Cart

Rat Monoclonal Antibody

CXCR5 (CD185) Monoclonal antibody specifically detects CXCR5 (CD185) in Human samples. It is validated for Flow Cytometry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CXCR5 (CD185)
Applications Flow Cytometry
Classification Monoclonal
Clone RF8B2
Conjugate Brilliant Blue 700 (BB700)
Host Species Rat
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Refrigerate (2 to 8°C)
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.