Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CXCR6 Antibody (1010910), PE/Atto594, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB6991PEATT594
Description
CXCR6 Monoclonal antibody specifically detects CXCR6 in Human samples. It is validated for Flow CytometrySpecifications
CXCR6 | |
Monoclonal | |
PE/ATTO 594 | |
PBS | |
Mouse | |
Protein A or G purified from ascites | |
RUO | |
Primary | |
Human | |
Purified |
Flow Cytometry | |
1010910 | |
Flow Cytometry | |
BONZOG-protein coupled receptor bonzo, CD186 antigen, CDw186, chemokine (C-X-C motif) receptor 6, C-X-C chemokine receptor type 6, CXC-R6, CXCR-6, G protein-coupled receptor, STRL33G-protein coupled receptor STRL33, TYMSTRCD186 | |
HEK293 human embryonic kidney cell line transfected with human CXCR6, Met1-Leu342, Accession # O00574 | |
0.1 mL | |
Chemokines and Cytokines, Cytokine Research, GPCR, Immunology, Innate Immunity, Virology Bacteria and Parasites | |
10663 | |
Store at 4C in the dark. Do not freeze. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction