Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CXCR6 Antibody (221002), DyLight 550, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals FAB2145L
Description
CXCR6 Monoclonal antibody specifically detects CXCR6 in Mouse samples. It is validated for Flow Cytometry, CyTOFSpecifications
CXCR6 | |
Monoclonal | |
DyLight 550 | |
50mM Sodium Borate | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Primary | |
Mouse | |
Purified |
Flow Cytometry, CyTOF | |
221002 | |
Flow Cytometry, CyTOF-ready | |
BONZOG-protein coupled receptor bonzo, CD186 antigen, CDw186, chemokine (C-X-C motif) receptor 6, C-X-C chemokine receptor type 6, CXC-R6, CXCR-6, G protein-coupled receptor, STRL33G-protein coupled receptor STRL33, TYMSTRCD186 | |
CHO Chinese hamster ovary cell line transfected with mouse CXCR6, Met1-Leu351, Accession # Q9EQ16 | |
0.1 mL | |
Chemokines and Cytokines, Cytokine Research, GPCR, Immunology, Innate Immunity, Virology Bacteria and Parasites | |
10663 | |
Store at 4°C in the dark. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction