Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CXCR6 (CD186) Mouse anti-Human, Brilliant Violet 786, Clone: 13B 1E5, BD Optibuild™

Catalog No. BDB743602 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB743602 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB743602 Supplier BD Biosciences Supplier No. 743602
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The 13B 1E5 monoclonal antibody specifically binds to C-X-C chemokine receptor type 6 (CXCR6), which is also known as CD186, BONZO, T-lymphocyte-expressed seven-transmembrane domain receptor (TYMSTR), and Seven transmembrane receptor-like from clone 33 (STRL33). CXCR6 is a G-protein coupled chemokine receptor that is expressed on subsets of activated and memory T cells and NKT cells. CXCR6 binds to soluble CXCL16 and membrane-anchored CXCL16 expressed by dendritic cells and macrophages. The CXCL16 and CXCR6 interaction activates Akt and mTor signaling. This regulates cellular migration including the recruitment of tumor-infiltrating lymphocytes and may contribute to the progression and metastasis of various cancers. CXCR6 also can serve as a coreceptor for certain strains of HIV-1 and HIV-2.

The antibody was conjugated to BD Horizon™ BV786 which is part of the BD Horizon Brilliant™ Violet family of dyes. This dye is a tandem fluorochrome of BD Horizon BV421 with an Ex Max of 405nm and an acceptor dye with an Em Max at 786nm. BD Horizon BV786 can be excited by the violet laser and detected in a filter used to detect Cy™7-like dyes (eg, 780/60nm filter). BV786 is a tandem fluorochrome of BD Horizon BV421 and an acceptor dye with an emission maximum at 786 nm. BV786 offers a bright choice for the 6th detector off the violet laser.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CXCR6 (CD186)
Classification Monoclonal
Clone 13B 1E5
Concentration 0.2mg/mL
Conjugate Brilliant Violet 786
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Gene Alias C-X-C chemokine receptor type 6, BONZO, STRL33, TYMSTR
Host Species Mouse
Immunogen Human CXCR6
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.