Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CXCR6 (CD186) Mouse anti-Human, BV421, Clone: 13B 1E5, BD Horizon™
Mouse Monoclonal Antibody
Supplier: BD Biosciences 566008
Description
CXCR6 (CD186) Monoclonal antibody specifically detects CXCR6 (CD186) in Human samples. It is validated for Flow cytometry.
CXCR6 (CD186) Monoclonal antibody specifically detects CXCR6 (CD186) in Human samples. It is validated for Flow cytometry.
Specifications
CXCR6 (CD186) | |
Monoclonal | |
0.2mg/mL | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
Mouse | |
25 μg | |
Cell Biology | |
Human | |
Purified |
Flow Cytometry | |
13B 1E5 | |
Brilliant Violet 421 | |
C-X-C chemokine receptor type 6; BONZO; STRL33; TYMSTR | |
Human CXCR6 | |
RUO | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with BD Horizon™ BV421 under optimum conditions and unconjugated antibody and free BD Horizon™ BV421 were removed. | |
IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only.
Spot an opportunity for improvement?Share a Content Correction