Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CXCR6 Monoclonal Antibody (2A4D10)
GREENER_CHOICE

Catalog No. FERMA548699
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FERMA548699 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FERMA548699 Supplier Invitrogen™ Supplier No. MA548699
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CXCR6 Monoclonal Antibody for Western Blot, Flow, ELISA

CD186, also known as CXCR6, is a G protein-coupled chemokine receptor reported to be one of the cellular receptors of HIV and SIV. CD186 is expressed by NKT cells, various subtypes of T cells with activated and memory phenotypes, and some plasma cells. It has been reported that mouse liver NK cells, but not naïve or splenic NK cells, express CD186. The ligand of CD186, CXCL16, is expressed by macrophages, dendritic cells and liver endothelial cells. Being highly expressed on NKT cells, CD186 is often used as a proxy marker of this cell subset.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CXCR6
Applications ELISA, Flow Cytometry, Western Blot
Classification Monoclonal
Clone 2A4D10
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% sodium azide
Gene CXCR6
Gene Accession No. O00574
Gene Alias BB217514; BONZO; C Cmotif chemokine; C X C motif chemokine; CC motif chemokine; CCmotif chemokine; CD186; CD186 antigen; CDw186; chemokine (C-X-C motif) receptor 6; chemokine receptor CXCR6; CXC; C-X-C chemokine receptor type 6; CXC motif chemokine; C-X-C motif chemokine receptor 6; Cxcr6; CXC-R6; CXCR-6; G protein-coupled receptor; G-protein coupled receptor bonzo; G-protein coupled receptor STRL33; OTTHUMP00000209997; OTTHUMP00000209998; STRL33; TYMSTR
Gene Symbols CXCR6
Host Species Mouse
Immunogen Purified recombinant fragment of human CD186 expressed in E. Coli.
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10663
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.