Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CXCR7/RDC-1 Antibody, mFluor Violet 500 SE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP224779MFV500
Description
CXCR7/RDC-1 Polyclonal antibody specifically detects CXCR7/RDC-1 in Human, Mouse, Rat, Sheep samples. It is validated for Flow CytometrySpecifications
CXCR7/RDC-1 | |
Polyclonal | |
Flow Cytometry | |
chemokine (C-X-C motif) receptor 7, Chemokine orphan receptor 1CMKOR1C-X-C chemokine receptor type 7, CXC-R7, CXCR-7, G protein-coupled receptor, GPR159G-protein coupled receptor 159, RDC-1, RDC1G-protein coupled receptor RDC1 homolog | |
A synthetic peptide corresponding amino acids 106-129 of human CXCR7/RDC1 was used as the immunogen, GenBank NP_064707.1. | |
0.1 mL | |
Chemokines and Cytokines, GPCR | |
57007 | |
Store at 4C in the dark. | |
IgG |
Flow Cytometry | |
mFluor Violet 500 SE | |
50mM Sodium Borate | |
Rabbit | |
Peptide affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat, Sheep | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction