Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CXCR7 Recombinant Rabbit Monoclonal Antibody (8V8E1)
GREENER_CHOICE

Catalog No. PIMA542902
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA542902 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIMA542902 Supplier Invitrogen™ Supplier No. MA542902
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

Positive test controls include: U-937, SH-SY5Y, HUVEC. Immunogen sequence: VCWLPYHVAV LLDIFSILHY IPFTCRLEHA LFTALHVTQC LSLVHCCVNP VLYSFINRNY RYELMKAFIF KYSAKTGLTK LIDASRVSET EYSALEQSTK.

This gene encodes a member of the G-protein coupled receptor family. Although this protein was earlier thought to be a receptor for vasoactive intestinal peptide (VIP), it is now considered to be an orphan receptor, in that its endogenous ligand has not been identified. The protein is also a coreceptor for human immunodeficiency viruses (HIV). Translocations involving this gene and HMGA2 on chromosome 12 have been observed in lipomas.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CXCR7
Applications ELISA, Western Blot
Classification Recombinant Monoclonal
Clone 8V8E1
Concentration 2 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene Ackr3
Gene Accession No. P25106
Gene Alias ACKR3; Atypical chemokine receptor 3; AW541270; C Cmotif chemokine; C X C motif chemokine; CC motif chemokine; CCmotif chemokine; chemokine; chemokine (C-X-C motif) receptor 7; chemokine orphan receptor 1; Cmkor1; CXC; C-X-C chemokine receptor type 7; CXC motif chemokine; CXCR7; CXC-R7; CXCR-7; G protein-coupled receptor; GPR159; G-protein coupled receptor 159; G-protein coupled receptor RDC1 homolog; RDC1; RDC-1
Gene Symbols Ackr3
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 263-362 of human CXCR7 (NP_0647071).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57007
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.