Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CXCR7 Recombinant Rabbit Monoclonal Antibody (HL2189)
GREENER_CHOICE

Catalog No. FEMA550423
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEMA550423 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEMA550423 Supplier Invitrogen™ Supplier No. MA550423
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

CXCR7 Recombinant Monoclonal Antibody for Western Blot, IHC (P), ICC/IF

This gene encodes a member of the G-protein coupled receptor family. Although this protein was earlier thought to be a receptor for vasoactive intestinal peptide (VIP), it is now considered to be an orphan receptor, in that its endogenous ligand has not been identified. The protein is also a coreceptor for human immunodeficiency viruses (HIV). Translocations involving this gene and HMGA2 on chromosome 12 have been observed in lipomas.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CXCR7
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone HL2189
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene Ackr3
Gene Accession No. O89039, P25106, P56485
Gene Alias ACKR3; Atypical chemokine receptor 3; AW541270; C Cmotif chemokine; C X C motif chemokine; CC motif chemokine; CCmotif chemokine; chemokine; chemokine (C-X-C motif) receptor 7; chemokine orphan receptor 1; Cmkor1; CXC; C-X-C chemokine receptor type 7; CXC motif chemokine; CXCR7; CXC-R7; CXCR-7; G protein-coupled receptor; GPR159; G-protein coupled receptor 159; G-protein coupled receptor RDC1 homolog; RDC1; RDC-1
Gene Symbols Ackr3
Host Species Rabbit
Immunogen Synthetic peptide of human CXCR7.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12778, 57007, 84348
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.